Merck
All Photos(1)
Z173479
Sigma-Aldrich
Double-ended filter with a glass stopcock on each of the two side-arms
capacity 60 mL, male Joints: ST/NS 14/20 (2)
NACRES:
NB.43
ラベル
BB
ジョイント
male Joints: ST/NS14/20 (2)
キャパシティ
60 mL
ポアサイズ
25-50 μm porosity
Related Categories
Compare Similar Items
View Full Comparison
1 of 4
This Item
Z173185Z220140SYNF973024M
label
BB
label
X
label
Y
label
-
joint
male Joints: ST/NS14/20 (2)
joint
female joint: ST/NS14/20
joint
joint ST/NS14/20 F, joint: ST/NS14/20 M
joint
joint: ST/NS24/40
capacity
60 mL
capacity
75 mL
capacity
100 mL
capacity
-
pore size
25-50 μm porosity
pore size
25-50 μm porosity
pore size
25-50 μm porosity
pore size
10-15 μm
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service