で組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|
この商品について
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Fortuna Graf 101.107-33
Volume:
1-5 mL
Feature:
subdivision 0.1 mL
Fitting:
GL 40, GL 45
現在、価格および在庫状況を閲覧できません。
feature
subdivision 0.1 mL
manufacturer/tradename
Fortuna Graf 101.107-33
volume measuring range
1-5 mL
volume
1-5 mL
fitting
GL 40, GL 45
General description
For dispensing strong acids and aggressive chemicals
Features:
Features:
- PTFE encased dispensing piston with sealing lip at its end
- FIX adapter made of PTFE
- Valve block made of PTFE, no metal springs in the valve - no corrosion
- Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
- Ergonmic finger protection cap
- Fast and easy volume setting with additional fine adjustment
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
1 of 1
当該品目 | |||
|---|---|---|---|
| manufacturer/tradename Fortuna Graf 101.107-33 | manufacturer/tradename Fortuna Graf 101.107-27 | manufacturer/tradename Fortuna Graf 101.107-48 | manufacturer/tradename Fortuna Graf 101.107-44 |
| volume 1-5 mL | volume 0.4-2 mL | volume 10-50 mL | volume 6-30 mL |
| feature subdivision 0.1 mL | feature subdivision 0.1 mL | feature subdivision 1.0 mL | feature subdivision 0.5 mL |
| fitting GL 40, GL 45 | fitting GL 40, GL 45 | fitting GL 40, GL 45 | fitting GL 40, GL 45 |
| volume measuring range 1-5 mL | volume measuring range 0.4-2.0 mL | volume measuring range 10-50 mL | volume measuring range 6-30 mL |
Still not finding the right product?
Explore all of our products under FORTUNA® Optifix® Safety ボトルトップディスペンサー