表示を変更する
| 容量 | カタログ番号 | 在庫状況 | 単価 |
|---|---|---|---|
| 1 ea | カートの在庫状況を確認する | ¥6,560 |
この商品について
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
25 mL
Material:
borosilicate glass , round bottom flask, single-neck
Packaging:
pack of 1
¥6,560
カートの在庫状況を確認する
material
borosilicate glass , round bottom flask, single-neck
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9592-15
capacity
25 mL
General description
With cap, FETFE® O-ring, and septum. Threaded <ST/>14/10 joint.
Note: This product is part of a kit. Please refer to the Z510297 web page for additional information.
Note: This product is part of a kit. Please refer to the Z510297 web page for additional information.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
1 of 1
当該品目 | |||
|---|---|---|---|
| material borosilicate glass , single-neck , round bottom flask | material borosilicate glass , round bottom flask, single-neck | material glass (Clear), round bottom flask, three angled neck | material borosilicate glass , round bottom flask, three-neck |
| manufacturer/tradename Ace Glass 9592-15 | manufacturer/tradename Ace Glass 9592-04 | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich |
| capacity 25 mL | capacity 5 mL | capacity 100 mL | capacity 100 mL |
| packaging pack of 1 | packaging pack of 1 | packaging case of 1 | packaging pack of 1 |
| feature closure type Screw cap | feature closure type Screw cap | feature closure type Standard Taper Joint | feature closure type Standard Taper joint |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
グローバルトレードアイテム番号
| カタログ番号 | GTIN |
|---|---|
| Z278262-1EA | 04061826660065 |