Merck
Z281514
Scienceware® Vikem® pipette aid
pkg of 6 ea
NACRES:
NB.01
物質
vinyl bulb
包装
pkg of 6 ea
manufacturer/tradename
Bel-Art F37886-0000
容量
85 mL
関連するカテゴリー
類似品目との比較
比較内容をすべて表示
Show Differences
1 of 4
当該品目
Z136085Z420093Z420115
Scienceware® Vikem® pipette aid pkg of 6 ea
Z281514
Scienceware® Vikem® pipette aid
Bel-Bulb™ pipettor vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm
Z136085
Bel-Bulbピペッター
manufacturer/tradename
Bel-Art F37886-0000
manufacturer/tradename
Bel-Art F37887-0000
manufacturer/tradename
Bel-Art
manufacturer/tradename
Bel-Art
volume
85 mL
volume
-
volume
-
volume
-
packaging
pkg of 6 ea
packaging
-
packaging
pack of 1
packaging
pack of 1
詳細
By squeezing the 85 mL vinyl bulb and releasing, fluid is drawn up into the pipette. The pipette aid is then removed, and the pipette top is quickly covered with a finger to control and release fluid. The tapered PE chuck allows for quick removal when pipette is filled.
法的情報
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください ( T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
この製品を見ている人はこちらもチェック
Slide 1 of 1
1 of 1
Pipette bulb volume 1 mL
Sigma-Aldrich
Z509035
Pipette bulb
テクニカルサービス連絡先