この商品について
NACRES:
NB.35
UNSPSC Code:
41101702
Manufacturer/tradename:
Coorstek 60358
Capacity:
7 mL
Material:
alumina
material
alumina
manufacturer/tradename
Coorstek 60358
O.D. × H
50 mm × 26 mm
capacity
7 mL
General description
99.5% alumina. Extremely hard and completely nonabsorbent surface. Completely glazed except for inside grinding surface. Order pestle separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
グローバルトレードアイテム番号
| カタログ番号 | GTIN |
|---|---|
| Z281999-1EA | 04061833309094 |
| Z282049-1EA | 04061833309117 |