Merck
Z406503
Stankovic transfer adapter
joint: ST/NS 24/40, thread size 20-400
ログイン組織・契約価格を表示する
サイズを選択してください
1 EA
¥9,360
¥9,360
単価¥12,480割引25%
出荷予定日2025年4月02日
サイズを選択してください
表示を変更する
1 EA
¥9,360
About This Item
UNSPSCコード:
41121815
NACRES:
NB.35
¥9,360
単価¥12,480割引25%
出荷予定日2025年4月02日
スレッドサイズ
20-400
ジョイント
joint: ST/NS24/40
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
These unique adapters greatly simplify the transfer of solids contained in round-bottom flasks into vials. Precision machined, chemically inert PTFE adapters will not seize in the joint. A wide range of thread sizes are available to accommodate most sample vials including scintillation vials (22 mm threads).
  • Transfers samples without exposure to air or moisture
  • Reduces sample losses due to air currents and static charge that can normally cause light solids to float or blow away when transferred open to the air
  • Excellent for transferring fluffy lyophilized samples, especially peptides
  • Transfers any freely flowing solid and eliminates weighing paper or other intermediate devices
Easy to Use:
Screw sample vial into bottle thread at top of adapter. Insert other end of adapter into flask joint. Invert assembly and gently tap vial on a soft surface to transfer solids from flask into sample vial.
その他情報
Care must be used when tapping vial to prevent accidental breakage. Tapping on a cork ring or other soft surface is recommended.
最新バージョンのいずれかを選択してください:
試験成績書(COA)