Z670359
Whatman® Nytran SuPerCharge (SPC) ナイロンブロッティングメンブレン
0.45 μm nylon blotting membranes 25 x 25 cm sheets, 10/pk
別名:
ナイロン ブロッティングメンブレン, ブロッティング膜
物質
Nylon membrane (Nytran SuPerCharge (SPC))
包装
pack of 10
manufacturer/tradename
Whatman 10416263
L × W
25 cm × 25 cm
ポアサイズ
0.45 μm
適合性
suitable for (Southern and Northern blotting; colony and plaque lifts and Dot-/Slot-Blots)
類似した製品をお探しですか? Visit 製品比較ガイド
詳細
Nytran SPC membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling.
With typical manufacturing techniques, increasing positive charge tends to increase background. Nytran SPC membranes are manufactured using a process that allows the combination of high positive charge with low background. Whether using radioactive or nonradioactive detection techniques, Nytran SPC consistently gives high signal with extremely low background.
  • Excellent signal-to-noise ratio
  • High positive charge
  • Consistent membrane morphology
  • High sensitivity
  • Flat, non-curling membrane
  • Nytran SuPerCharge binding capacity: > 600 μg/cm2
法的情報
Nytran is a trademark of Cytiva
Whatman is a registered trademark of Cytiva