These GL 45 connection system caps are suitable for all bottles with a GL 45 thread. Please see the links below to review several suitable bottle options available:
https://www.sigmaaldrich.com/product/aldrich/z742009
https://www.sigmaaldrich.com/product/aldrich/z742012
https://www.sigmaaldrich.com/product/aldrich/z742008
Z680397
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 2 x GL 14 ports
別名:
1129750, Duran Cap Accessories, GL 45 Connection Caps
ログイン組織・契約価格を表示する
サイズを選択してください
2 EA
¥27,700
¥27,700
Check Cart for Availability
すべての画像(1)
サイズを選択してください
表示を変更する
2 EA
¥27,700
About This Item
UNSPSCコード:
41121800
NACRES:
NB.17
¥27,700
Check Cart for Availability
おすすめの製品
物質
blue-gray polypropylene
メーカー/製品名
DURAN® 1129750
パラメーター
-40-140 °C temp. limit
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Sterile pressure equalization is possible through use of membrane filter.
特徴および利点
- Manufactured from Polypropylene.
- Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts.
- Available with 2, or 3 threaded outlets (ports).
- Four different tubing diameters (1.6 mm, 3.0 mm, 3.2 mm or 6.0 mm tubing outer diameter) can be used by changing the GL 14 inserts.
- By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap.
- Fully compatible with all the GL 45 threads and therefore highly versatile.
法的情報
DURAN is a registered trademark of DWK Life Sciences
最新バージョンのいずれかを選択してください:
この製品を見ている人はこちらもチェック
-
Hi, What bottle would be best to use with cap GL 45 Z680397 with 6 mm diameter ports? I am looking to use a bottle of 250 to 500 ml. Thank you
1 回答-
役に立ちましたか?
-
-
Hi, I am wondering if all the following parts are included in your product. Insert for screw cap GL 14, 3.0 mm (2 pcs per one cap) and Screw Cap GL 14 Hose Connection (2 pcs per one cap). Thanks.
1 回答-
This product is a GL45 screw cap with two ports that have GL14 threads as shown in product image. The inserts and hose connections are not included, but are sold separately.