glass (Type 1/ neutral glass)
graduations
DURAN®
Schott
5000 mL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | Z680915 | Z680907 | Z680885 |
---|---|---|---|
material glass (Type 1/ neutral glass) | material glass (Type 1/ neutral glass) | material glass (Type 1/ neutral glass) | material glass (Type 1/ neutral glass) |
feature graduations | feature graduations | feature graduations | feature graduations |
capacity 5000 mL | capacity 600 mL | capacity 400 mL | capacity 150 mL |
manufacturer/tradename DURAN® | manufacturer/tradename DURAN®, Schott | manufacturer/tradename DURAN®, Schott | manufacturer/tradename DURAN®, Schott |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service