Z691518
MagLab™ 保存システム
starter pack
ログイン組織・契約価格を表示する
サイズを選択してください
4 EA
¥11,960
¥11,960
カートの在庫状況を確認する
サイズを選択してください
表示を変更する
4 EA
¥11,960
About This Item
UNSPSCコード:
41122800
NACRES:
NB.35
¥11,960
カートの在庫状況を確認する
メーカー/製品名
Heathrow Scientific HD23500
L × W × H
522 mm × 393 mm × 102 mm
色
assorted colors
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | Z691542 | Z691550 | Z691534 |
---|---|---|---|
manufacturer/tradename Heathrow Scientific HD23500 | manufacturer/tradename Heathrow Scientific HD23503 | manufacturer/tradename Heathrow Scientific HD23504 | manufacturer/tradename Heathrow Scientific HD23502 |
L × W × H 522 mm × 393 mm × 102 mm | L × W × H 131 mm × 112 mm × 76 mm | L × W × H 130 mm × 78 mm × 102 mm | L × W × H 130 mm × 91 mm × 98 mm |
color assorted colors | color purple | color orange | color green |
詳細
With lab bench top space at a premium, common lab products such as pipettors, test tubes and miscellanous accessories can be easily organized with Heathrow Scientific′s magnetic lab
storage system, the MagLab. MagLab includes four magnetic pieces: “MagRack” that holds (9) 15ml and (4) 50ml tubes; “Mag Wipe” that holds a standard size (11cm x 21cm) Kimwipes; box; “MagPette” that
holds two pipettors (universal design fits most common brands); and “MagBox” that holds miscellaneous items such as pens/pencils, notes, etc. Each rack is made of robust lightweight ABS plastic in bright and
fun translucent colors and features four powerful magnets for a secure hold onto most metal surfaces or create your own holding point with the metal plate included for each segment.
storage system, the MagLab. MagLab includes four magnetic pieces: “MagRack” that holds (9) 15ml and (4) 50ml tubes; “Mag Wipe” that holds a standard size (11cm x 21cm) Kimwipes; box; “MagPette” that
holds two pipettors (universal design fits most common brands); and “MagBox” that holds miscellaneous items such as pens/pencils, notes, etc. Each rack is made of robust lightweight ABS plastic in bright and
fun translucent colors and features four powerful magnets for a secure hold onto most metal surfaces or create your own holding point with the metal plate included for each segment.
法的情報
MagLab is a trademark of Heathrow Scientific
最新バージョンのいずれかを選択してください:
アクティブなフィルタ