Z742077

Exact-Temp Coated bottle thermometer with 2 point DAkkS-Calibration

for incubator, temperature +18 to +50 °C, precision: ±0.5 °C

NACRES:
NB.79

物質

bottle ( Ethylene/glycol)

詳細

I-030-1S

特徴

accuracy± 1 °C
calibration (2 point DAkkS)
safety coating

manufacturer/tradename

80 001 15/04/2P (Ludwig Schneider)

temperature measuring range

+18 to +50 °C, precision: ±0.5 °C

L

130 mm

red

類似した製品をお探しですか? Visit 製品比較ガイド

詳細

Exact-temp thermometers with DAkkS-Calibration are high-precise instruments to monitor cooling and tempering devices in laboratories, medical technology and R&D.
Each thermometer is mounted in an unbreakable plastic bottle, filled with a special medium to ensure accurate measurings even at open doors.
With DAkkS-Calibration certificate (two calibration points as standard) these high precision thermometers are especially suitable for monitoring certified quality management systems.
They are supplied with magnets for spacesaving wall or door fixing. Exact-temp thermometers are very reliable and economical because they work independent from any mains or battery connection.
Traceable to international standards Exact-temp thermometers are available with organic filling in varying measuring ranges and scale units.
The instruments are carefully aged to ensure a high accuracy. Exact-temp thermometers are calibrated with a reference and are therefore traceable to international standards.
試験成績書(COA)
原産地証明書(COO)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。