Merck
  • 324833M
All Photos(1)

324833-M

Millipore

EGFR Inhibitor III

The EGFR Inhibitor III, also referenced under CAS 733009-42-2, controls the biological activity of EGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):
EGFR Inhibitor III, N-(4-((3,4-Dichloro-6-fluorophenyl)amino)-quinazolin-6-yl)-2-chloroacetamide
Empirical Formula (Hill Notation):
C16H10Cl3FN4O
CAS Number:
Molecular Weight:
399.63

アッセイ

≥98% (HPLC)

形状

solid

manufacturer/tradename

Calbiochem®

保管条件

OK to freeze
protect from light

yellow

溶解性

DMSO: 100 mg/mL (Use fresh, anhydrous DMSO.)

保管温度

−20°C

Compare Similar Items

View Full Comparison

1 of 4

This Item
324832539652324732
EGFR Inhibitor III The EGFR Inhibitor III, also referenced under CAS 733009-42-2, controls the biological activity of EGFR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Millipore

324833-M

EGFR Inhibitor III

ErbB2 Inhibitor II The ErbB2 Inhibitor II, also referenced under CAS 928207-02-7, controls the biological activity of ErbB2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sigma-Aldrich

324732

ErbB2 Inhibitor II

assay

≥98% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

color

yellow

color

yellow

color

orange

color

yellow

solubility

DMSO: 100 mg/mL (Use fresh, anhydrous DMSO.)

solubility

water: 1 mg/mL, DMSO: 25 mg/mL

solubility

DMSO: 20 mg/mL

solubility

DMSO: 50 mg/mL

詳細

A cell-permeable quinazolinyl-chloroacetamide compound that selectively and irreversibly blocks EGFR autophosphorylation with no effect on the expression levels of EGFR, and suppresses leiomyoma and myometrical cell proliferation (IC50 = 0.7 µM and 1.1 µM, respectively). Shown to induce apoptosis and cell cycle arrest, and in combination with Genistein (Cat. No. 345834) completely suppress leiomyoma cell proliferation.
A cell-permeable quinazolinyl-chloroacetamide compound that selectively and irreversibly blocks EGFR autophosphorylation with no effect on the expression levels of EGFR, and suppresses leiomyoma and myometrical cell proliferation (IC50 = 0.7 µM and 1.1 µM, respectively). Shown to induce apoptosis and cell cycle arrest, and in combination with Genistein (Cat. No. 345834) completely suppress leiomyoma cell proliferation.

生物化学的/生理学的作用

Primary Target
EGFR
Target IC50: 0.7 µM and 1.1 µM suppressesing leiomyoma and myometrical cell proliferation, respectively

警告

Toxicity: Irritant (B)

再構成

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

その他情報

Shushan, A., et al. 2007. Fertil. Steril.87, 127.

法的情報

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

保管分類コード

11 - Combustible Solids

WGK

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

(enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service