Merck
All Photos(1)
516557
Sigma-Aldrich
Anti-PGC-1, C-Terminal (777-797) Rabbit pAb
liquid, Calbiochem®
Synonym(s):
Anti-Ligand Effect Modulator 6, Anti-PPARγ Coactivator 1
由来生物
rabbit
品質水準
100
抗体製品の状態
affinity isolated antibody
antibody product type
primary antibodies
クローン
polyclonal
形態
liquid
species reactivity
human, mouse
manufacturer/tradename
Calbiochem®
保管条件
OK to freeze
avoid repeated freeze/thaw cycles
アイソタイプ
IgG
輸送温度
wet ice
保管温度
−70°C
Gene Information
詳細
Recognize the ~92-105 kDa (apparent MW) PGC-1 protein.
Immunoaffinity purified rabbit polyclonal antibody. Recognize the ~92-105 kDa (apparent MW) PGC-1 protein.
This Anti-PGC-1, C-Terminal (777-797) Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunohistochemistry for the detection of PGC-1, C-Terminal (777-797).
免疫原
Human
a synthetic peptide [(C)SKYDSLDFDSLLKEAQRSLRR] corresponding to amino acids 777-797 of the human PPARγ Coactivator 1 protein
包装
100 μL in Plastic ampoule
アプリケーション
ELISA (1:50,000)
Immunoblotting (1:1000)
Immunohistochemistry (1:300)
警告
Toxicity: Standard Handling (A)
物理的形状
In PBS.
再構成
Following initial thaw, aliquot and freeze (-70°C).
その他情報
Puigserver, P., et al. 1998. Cell 92, 829.
Variables associated with assay conditions will dictate the optimal working dilution.
法的情報
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
保管分類コード
12 - Non Combustible Liquids
WGK
nwg
Not applicable
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Quality
Enter Lot Number to search for Certificate of Quality (COQ).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service