MAB13439

Sigma-Aldrich

Anti-MMP-1 Antibody, clone 4E7.2

clone 4E7.2, Chemicon®, from mouse

別名:
Fibroblast Collagenase, Interstitial Collagenase
eCl@ss:
32160702

品質水準

100

由来生物

mouse

antibody product type

primary antibodies

クローン

4E7.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

アプリケーション

ELISA: suitable
western blot: suitable

アイソタイプ

IgG1

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

dry ice

特異性

Antibody MAB13439 is directed against a synthetic peptide corresponding to the amino terminal region of human MMP-1. The antibody binds to MMP-1 but does not cross-react with the other MMP family members (MMP-2, MMP-3, MMP-9, etc.).

Reacts with human. Other species not tested; however this sequence of MMP-1 is well conserved across species. Higher concentrations of antibody may be needed for non-human samples.

アプリケーション

Western blot: 1:1000 when using colorimetric substrates; 1:5,000 for chemiluminescent substrates. When used against the reduced protein, antibody MAB13439 recognizes bands at 53 kDa and 51 kDa (the pro-form), as well as active forms.

ELISA

Collagenase levels in quiescent cells and tissues is minimal, and stimulation or protein concentration is often needed to visualize the bands. In addition, cell types differ greatly in the quantity of collagenase produced.

Optimal working dilutions must be determined by end user.
This Anti-MMP-1 Antibody, clone 4E7.2 is validated for use in ELISA, WB for the detection of MMP-1.
Research Category
細胞骨格
Research Sub Category
MMP及び組織メタロプロテアーゼ

関連事項

Replaces: 04-1122

物理的形状

Purified immunoglobulin. Liquid at 1 mg/mL containing 0.1% sodium azide.
Format: Purified

保管および安定性

Maintain at -20°C in undiluted aliquots for up to 12 months.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

その他情報

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

法的情報

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

ドイツ水質汚染分類

WGK 1

引火点(°F)

Not applicable

引火点(℃)

Not applicable

Yi Wang et al.
Molecular medicine reports, 12(3), 4389-4395 (2015-07-02)
Subacromial bursitis (SAB) is the major source of pain in rotator cuff disease. Although multiple investigations have provided support for the role of inflammatory cytokines in SAB, few have focussed on the use these cytokines in the treatment of SAB....
Ta-Kuo Juan et al.
Oncology letters, 15(5), 7716-7724 (2018-05-08)
Tetrandrine (TET) exhibits biological activities, including anticancer activity. In Chinese medicine, TET has been used to treat hypertensive and arrhythmic conditions and has been demonstrated to induce cytotoxic effects on human cancer cell lines. However, to the best of the...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。