QCMIC205
Millipore
Total & Fecal Streptococcus/Enterococcus - WP QC 4 Levels
グレード
certified reference material
包装
set of
manufacturer/tradename
RTC QC-MIC-205
主な業界
Environmental
Food and Beverages
出荷済み
wet ice
保管温度
2-8°C
適合性
Enterococcus spp.
Streptococcus spp.
詳細
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
包装
Set contains 8 sterile screw cap vials each containing a Vitroid (a small disk containing the organisms) and 8 - 99mL bottles of phosphate buffer.
その他情報
Refer to the Certificate of Analysis for additional information.
法的情報
Vitroid is a trademark of Sigma-Aldrich Co. LLC
RIDADR
UN3373 - class 6.2 Biological substance, Category B
ドイツ水質汚染分類
3