サイズを選択してください
| あなたへ/カタログ番号 | 在庫状況 | 単価 |
|---|---|---|
1 ea | カートの在庫状況を確認する | ¥26,400 |
この商品について
¥26,400
material
borosilicate glass , round bottom flask, three-neck (Vertical), three-neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
Legal Information
1 of 1
当該品目 | |||
|---|---|---|---|
| manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Sigma-Aldrich |
| capacity 100 mL | capacity 100 mL | capacity 1,000 mL | capacity 1,000 mL |
| material borosilicate glass , three-neck (Vertical), round bottom flask | material borosilicate glass , round bottom flask, three-neck (Vertical) | material borosilicate glass , round bottom flask, three-neck (Vertical) | material borosilicate glass , round bottom flask, three-neck (Vertical) |
| joint joint: ST/NS24/40 | joint joint: ST/NS14/20 | joint joint: ST/NS29/32 | joint joint: ST/NS24/40 |
| packaging pack of 1 | packaging pack of 1 | packaging pack of 1 | packaging pack of 1 |
| feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint | feature closure type Standard Taper Joint |
Still not finding the right product?
Explore all of our products under Aldrich® three-neck round-bottom flask