Merck
HS120791
Lab Drying Rack
white polystyrene (blue & green pegs)
ログイン組織・契約価格を表示する
サイズを選択してください
1 EACH
¥34,440
¥34,440
出荷予定日2025年4月10日
サイズを選択してください
表示を変更する
1 EACH
¥34,440
About This Item
UNSPSCコード:
41122800
NACRES:
NB.12
¥34,440
出荷予定日2025年4月10日
物質
white polystyrene (blue & green pegs)
特徴
to hold 36 pieces small glassware plus 36 pieces large glassware
包装
pkg of 1 ea
メーカー/製品名
Heathrow Scientific 120791
L × W × H
441 mm × 660 × 139 mm
詳細
Included:-72 Pegs ∼ 36 Small (Green) ∼ 36 Large (Blue)-2 Drying Baskets-Drainage Tubing-Mounting Hardware
特徴および利点
  • Most Versatile Rack in the market
  • Channels designed for more useful configurations
  • Large and Small pegs for all glassware varieties
  • One of a kind Drying Basket to collect smaller items
  • Designed to protect against breakage
  • Channels designed for more useful configurations
  • Peg angle allows for better hold - especially large, heavy pieces Patent Pending SureGrip Hook securely grips glassware into place
最新バージョンのいずれかを選択してください:
試験成績書(COA)