SAB1402586

Sigma-Aldrich

Monoclonal Anti-CMPK antibody produced in mouse

clone 1D7, purified immunoglobulin, buffered aqueous solution

別名:
UMP-CMPK, CMPK, UMPK, UMK, CMK
MDL番号:
NACRES:
NA.41

品質水準

100

由来生物

mouse

抗体製品の状態

purified immunoglobulin

antibody product type

primary antibodies

クローン

1D7, monoclonal

形態

buffered aqueous solution

分子量

antigen ~51.19 kDa

species reactivity

human

アプリケーション

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

アイソタイプ

IgG2aκ

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

dry ice

保管温度

−20°C

Gene Information

human ... CMPK1(51727)

詳細

Uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) catalyzes the phosphoryl transfer from ATP to UMP, CMP, and deoxy-CMP (dCMP), resulting in the formation of ADP and the corresponding nucleoside diphosphate. These nucleoside diphosphates are required for cellular nucleic acid synthesis (Liou et al., 2002 [PubMed 11912132]).[supplied by OMIM

免疫原

CMPK (AAH14961, 1 a.a. ~ 228 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLSRCRSRLLHVLGLSFLLQTRRPILLCSPRLMKPLVVFVLGGPGAGKGTQCARIVEKYGYTHLSAGELLRDERKNPDSQYGELIEKYIKEGKIVPVEITISLLKREMDQTMAANAQKNKFLIDGFPRNQDNLQGWNKTMDGKADVSFVLFFDCNNEICIERCLERGKSSGRSDDNRESLEKRIQTYLQSTKPIIDLYEEMGKVKKIDASKSVDEVFDEVVQIFDKEG

物理的形状

無色透明のPBS溶液、pH 7.4

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Aiko Nobusawa et al.
Pathology international, 64(1), 20-27 (2014-01-30)
Diagnosis of the exact grade of oral epithelial dysplasia is difficult, and interobserver variations in grading are common. The aim of this study was to investigate the expression patterns of cytokeratins (CKs) in dysplastic oral epithelia, to identify useful double...
T Nowikiewicz et al.
Neoplasma, 61(3), 299-304 (2014-05-16)
According to current therapeutic guidelines, finding micrometastases in the sentinel node (SLN) of a patient with non-advanced breast cancer is not an absolute indication for adjuvant axillary lymph node dissection (ALND). This work presents our experiences regarding this clinical problem....
Florencia McAllister et al.
Cancer biology & therapy, 15(6), 688-698 (2014-03-13)
Deoxycytidine kinase (dCK) and human antigen R (HuR) have been associated with response to gemcitabine in small studies. The present study investigates the prognostic and predictive value of dCK and HuR expression levels for sensitivity to gemcitabine and 5-fluorouracil (5-FU)...
Majid Ebrahimi Warkiani et al.
The Analyst, 139(13), 3245-3255 (2014-05-21)
The detection and characterization of rare circulating tumor cells (CTCs) from the blood of cancer patients can potentially provide critical insights into tumor biology and hold great promise for cancer management. The ability to collect a large number of viable...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。