SAB1411851

Sigma-Aldrich

ANTI-ADIPOQ antibody produced in mouse

clone 4H2, purified immunoglobulin, buffered aqueous solution

別名:
ACDC, APM-1
NACRES:
NA.41

品質水準

100

由来生物

mouse

抗体製品の状態

purified immunoglobulin

antibody product type

primary antibodies

クローン

4H2, monoclonal

形態

buffered aqueous solution

species reactivity

human

アプリケーション

immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

アイソタイプ

IgG2bκ

結合体

unconjugated

UniProtアクセッション番号

出荷済み

dry ice

保管温度

−20°C

Gene Information

human ... ADIPOQ(9370)

免疫原

Recombinant Flag fusion protein corresponding to full length Adiponectin.
Mouse monoclonal antibody raised against adiponectin.

物理的形状

無色透明のPBS溶液、pH 7.4

RIDADR

NONH for all modes of transport

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Shen-Nien Wang et al.
Journal of gastroenterology, 49(9), 1342-1351 (2013-10-18)
Alterations of adiponectin (APN), one of the adipokines, have been associated with human cancers. However, the clinical significance and impacts of APN on hepatocellular carcinoma (HCC) remain undetermined. Using immunohistochemistry, expression patterns of APN were semiquantitatively scored and further statistically...
Ian R W Ritchie et al.
The Journal of physiology, 592(12), 2653-2665 (2014-04-02)
Adiponectin (Ad) has been proposed to be a regulator of mitochondrial biogenesis in skeletal muscle, and necessary for exercise-induced increases in mitochondrial content. We first confirmed that Ad could acutely increase the expression of mitochondrial proteins during a 10 h...
Tadateru Iwayama et al.
Journal of cardiology, 63(5), 344-349 (2013-11-16)
Epicardial adipose tissue (EAT) surrounding the heart may contribute to the development of coronary artery disease (CAD) through its local secretion of adipocytokines. Although the quantity of EAT is associated with obesity and metabolic syndrome, the role of EAT in...
Helene Baribault et al.
Expert opinion on therapeutic targets, 18(11), 1253-1264 (2014-10-08)
Although the human genome encodes ∼ 20,000 protein-coding genes, only a very small fraction of these have been explored as potential targets for therapeutic development. The challenge of identifying and validating new protein targets has contributed to the significant reduction...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。