SAB1412724

Sigma-Aldrich

Anti-KRT82 antibody produced in mouse

purified immunoglobulin

別名:
Anti-KRTHB2, Anti-HB2, Anti-Hb-2
NACRES:
NA.41

品質水準

100

由来生物

mouse

抗体製品の状態

purified immunoglobulin

antibody product type

primary antibodies

形態

buffered aqueous solution

分子量

antigen 56.43 kDa

species reactivity

human

アプリケーション

western blot: 1 μg/mL

結合体

unconjugated

GenBank® accession no.

UniProtアクセッション番号

出荷済み

dry ice

保管温度

−20°C

Gene Information

human ... KRT82(3888)

詳細

The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this keratin appears to be a hair cuticle-specific keratin. (provided by RefSeq)

免疫原

KRT82 (AAI53100.1, 1 a.a. ~ 513 a.a) full-length human protein.

Sequence
MSYHSFQPGSRCGSQSFSSYSAVMPRMVTHYAVSKGPCRPGGGRGLRALGCLGSRSLCNVGFGRPRVASRCGGTLPGFGYRLGATCGPSACITPVTINESLLVPLALEIDPTVQRVKRDEKEQIKCLNNRFASFINKVRFLEQKNKLLETKWNFMQQQRCCQTNIEPIFEGYISALRRQLDCVSGDRVRLESELCSLQAALEGYKKKYEEELSLRPCVENEFVALKKDVDTAFLMKADLETNAEALVQEIDFLKSLYEEEICLLQSQISETSVIVKMDNSRELDVDGIIAEIKAQYDDIASRSKAEAEAWYQCRYEELRVTAGNHCDNLRNRKNEILEMNKLIQRLQQETENVKAQRCKLEGAIAEAEQQGEAALNDAKCKLAGLEEALQKAKQDMACLLKEYQEVMNSKLGLDIEIATYRRLLEGEEHRLCEGIGPVNISVSSSKGAFLYEPCGVSTPVLSTGVLRSNGGCSIVGTGELYVPCEPQGLLSCGSGRKSSMTLGAGGSSPSHKH

特徴および利点

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

物理的形状

無色透明のPBS溶液、pH 7.4

法的情報

GenBank is a registered trademark of United States Department of Health and Human Services

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。