SAB2100460

Sigma-Aldrich

抗COG2抗体 ウサギ宿主抗体

affinity isolated antibody

別名:
抗LDLC抗体, 抗オリゴマーゴルジ複合体成分2抗体

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

83 kDa

species reactivity

rat, human, rabbit, mouse, horse

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... COG2(22796)

免疫原

Synthetic peptide directed towards the N terminal region of human COG2

生物化学的/生理学的作用

Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG2. Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG2 (Ungar et al., 2002 [PubMed 11980916]).[supplied by OMIM].

シーケンス

Synthetic peptide located within the following region: KRVQLEELRDDLELYYKLLKTAMVELINKDYADFVNLSTNLVGMDKALNQ

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。