SAB2102625

Sigma-Aldrich

抗UBE2L6抗体 ウサギ宿主抗体

affinity isolated antibody

別名:
抗MGC40331抗体, 抗RIG-B抗体, 抗ユビキチン結合酵素 E2L 6抗体, 抗UBCH8抗体
NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

18 kDa

species reactivity

rabbit, human

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... UBE2L6(9246)

免疫原

Synthetic peptide directed towards the N terminal region of human UBE2L6

生物化学的/生理学的作用

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is highly similar in primary structure to the enzyme encoded by UBE2L3 gene. The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is highly similar in primary structure to the enzyme encoded by UBE2L3 gene. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

シーケンス

Synthetic peptide located within the following region: LLLPDQPPYHLKAFNLRISFPPEYPFKPPMIKFTTKIYHPNVDENGQICL

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。