SAB2103597

Sigma-Aldrich

Anti-SULT1B1, (N-terminal) antibody produced in rabbit

affinity isolated antibody

別名:
抗SULT2B1抗体, 抗MGC13356抗体, 抗ST1B2抗体
NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

35 kDa

species reactivity

pig, human, dog

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... SULT1B1(27284)

免疫原

Synthetic peptide directed towards the N terminal region of human SULT1B1

生物化学的/生理学的作用

SULT1B1 Catalyzes the sulfate conjugation of many hormones, neurotransmitters, drugs and xenobiotic compounds. Sulfonation increases the water solubility of most compounds, and therefore their renal excretion, but it can also result in bioactivation to form active metabolites. Sulfates dopamine, small phenols such as 1-naphthol and p-nitrophenol and thyroid hormones, including 3,3′-diiodothyronine, triidothyronine, reverse triiodothyronine and thyroxine.Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes.

シーケンス

Synthetic peptide located within the following region: KRGFITEKVPMLEMTLPGLRTSGIEQLEKNPSPRIVKTHLPTDLLPKSFW

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。