SAB2105612

Sigma-Aldrich

Anti-ENPP2, (N-terminal) antibody produced in rabbit

affinity isolated antibody

別名:
Anti-ATX-X, Anti-FLJ26803, Anti-NPP2, Anti-ATX, Anti-LysoPLD
NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

95 kDa

species reactivity

mouse, guinea pig, human, horse, dog, rat, rabbit

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... ENPP2(5168)

免疫原

Synthetic peptide directed towards the N terminal region of human ENPP2

生物化学的/生理学的作用

The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5′ end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.

シーケンス

Synthetic peptide located within the following region: YTLATGLYPESHGIVGNSMYDPVFDATFHLRGREKFNHRWWGGQPLWITA

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Se Jin Park et al.
American journal of rhinology & allergy, 28(3), 199-207 (2014-07-02)
Chronic sinusitis with nasal polyps (CRSwNPs) or CRS without NPs (CRSsNPs) is associated with expression of various cytokines. Lysophosphatidic acid (LPA) generated by autotaxin (ATX), LPA-producing enzyme, initiates signaling cascade involved in the inflammatory responses and participates in diverse biological...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。