Merck
  • SAB2107504
All Photos(1)

SAB2107504

Sigma-Aldrich

Anti-OVGP1 antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

由来生物

rabbit

品質水準

結合体

unconjugated

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形状

buffered aqueous solution

分子量

75 kDa

species reactivity

human

濃度

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBIアクセッション番号

UniProtアクセッション番号

輸送温度

wet ice

保管温度

−20°C

Gene Information

human ... OVGP1(5016)

免疫原

Synthetic peptide directed towards the C terminal region of human OVGP1

シーケンス

Synthetic peptide located within the following region: QLPEQTPLAFDNRFVPIYGNHSSVNSVTPQTSPLSLKKEIPENSAVDEEA

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

保管分類コード

12 - Non Combustible Liquids

WGK

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service