SAB2107656

Sigma-Aldrich

Anti-CDK11A antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

84 kDa

species reactivity

horse, guinea pig, rat, mouse, rabbit, dog, human, bovine

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... CDK11A(728642)

詳細

The gene CDK11A (cyclin dependent kinase 11A) is mapped to human chromosome 1p36.33. Human genome contains two genes for CDK11, CDK11A and CDK11B. Both genes have three isoforms, the long CDK11p110, short CDK11p58 and CDK11p46. CDK11p110 is present in the nucleus, CDK11p58 is present in the nucleus and cytoplasm, and CDK11p46 is in the cytoplasm. CDK11p58 is mainly expressed in the G2/M phase. CDK11p110 is expressed during the whole cell cycle. The CDK11A protein has many NLSs (nuclear localization sequences), a 14-3-3 consensus site, an arginine/glutamic acid domain (RE domain), a poly-glutamic acid domain (poly-E domain) and a carboxy-terminal catalytic domain.

免疫原

The immunogen for anti-CDK11A antibody: synthetic peptide derected towards the C terminal of human CDK11A

生物化学的/生理学的作用

CDK11 (cyclin dependent kinase 11) regulates transcription and RNA processing (mainly alternate splicing) by binding to L-type cyclins. It is also involved in cellular responses, such as hormone receptor signaling and autophagy. CDK11p58 is needed for duplication of the centrioles, spindle rearrangement and sister chromatid cohesion at centromeres. CDK11 plays an important role in cancer cell growth and proliferation by regulating cell death.

シーケンス

Synthetic peptide located within the following region: EYFRETPLPIDPSMFPTWPAKSEQQRVKRGTSPRPPEGGLGYSQLGDDDL

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Clotilde Petretti et al.
EMBO reports, 7(4), 418-424 (2006-02-08)
The CDK11 (cyclin-dependent kinase 11) gene has an internal ribosome entry site (IRES), allowing the expression of two protein kinases. The longer 110-kDa isoform is expressed at constant levels during the cell cycle and the shorter 58-kDa isoform is expressed...
Marcos Malumbres
Genome biology, 15(6), 122-122 (2014-09-03)
Cyclin-dependent kinases (CDKs) are protein kinases characterized by needing a separate subunit - a cyclin - that provides domains essential for enzymatic activity. CDKs play important roles in the control of cell division and modulate transcription in response to several...
Yubing Zhou et al.
Oncotarget, 7(26), 40846-40859 (2016-10-27)
Overexpression and/or hyperactivation of cyclin-dependent kinases (CDKs) are common features of most cancer types. CDKs have been shown to play important roles in tumor cell proliferation and growth by controlling cell cycle, transcription, and RNA splicing. CDK4/6 inhibitor palbociclib has...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。