SAB2107773

Sigma-Aldrich

Anti-KLRK1 antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

24 kDa

species reactivity

human

濃度

0.5 mg - 1 mg/mL

アプリケーション

western blot: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... KLRK1(22914)

免疫原

The immunogen for anti-KLRK1 antibody: synthetic peptide derected towards the C terminal of human KLRK1

シーケンス

Synthetic peptide located within the following region: HIPTNGSWQWEDGSILSPNLLTIIEMQKGDCALYASSFKGYIENCSTPNT

物理的形状

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

ドイツ水質汚染分類

WGK 3

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Laleh Talebian et al.
Transfusion, 54(6), 1515-1521 (2014-01-23)
The NKG2D receptor, one of the natural killer (NK) cell-activating receptors, is expressed on the surface of CD3+CD8+ T cells, γδ+ T cells, NK cells, NKT cells, and a few CD4+ T cells. We show, for the first time, a...
Hongshuang Ma et al.
Clinical rheumatology, 33(11), 1603-1610 (2014-07-16)
Imbalance of natural killer (NK) cells is associated with the development of systemic lupus erythematosus (SLE). However, little is known about the dynamic changes on NK cells following therapy. This study aimed at examining the impact of classic therapies on...
Javier Celis-Gutierrez et al.
The EMBO journal, 33(17), 1928-1940 (2014-06-26)
Natural killer (NK) cells are involved in immune responses against tumors and microbes. NK-cell activation is regulated by intrinsic and extrinsic mechanisms that ensure NK tolerance and efficacy. Here, we show that the cytoplasmic signaling molecules Dok1 and Dok2 are...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。