SAB3500874

Sigma-Aldrich

Monoclonal Anti-HEMAGGLUTININ antibody produced in mouse

clone 4E10C10, affinity isolated antibody

NACRES:
NA.41

品質水準

100

由来生物

mouse

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

4E10C10, monoclonal

形態

buffered aqueous solution

species reactivity

virus

濃度

1 mg/mL

アプリケーション

ELISA: suitable
immunoblotting: suitable

結合体

unconjugated

NCBIアクセッション番号

出荷済み

wet ice

保管温度

−20°C

免疫原

Antibody was raised against recombinant protein corresponding to amino acids 17 - 338 of H5 HA1.

特徴および利点

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

物理的形状

1 mg/mLのPBS溶液(0.02%アジ化ナトリウム含有)

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
C Buxton Bridges et al.
The Journal of infectious diseases, 181(1), 344-348 (1999-12-23)
The first outbreak of avian influenza A (H5N1) occurred among humans in Hong Kong in 1997. To estimate the risk of person-to-person transmission, a retrospective cohort study was conducted to compare the prevalence of H5N1 antibody among health care workers...
K F Shortridge et al.
Virology, 252(2), 331-342 (1999-01-08)
The transmission of avian H5N1 influenza viruses to 18 humans in Hong Kong in 1997 with six deaths established that avian influenza viruses can transmit to and cause lethal infection in humans. This report characterizes the antigenic and biological properties...
D J Alexander
Veterinary microbiology, 74(1-2), 3-13 (2000-05-09)
Only type A influenza viruses are known to cause natural infections in birds, but viruses of all 15 haemagglutinin and all nine neuraminidase influenza A subtypes in the majority of possible combinations have been isolated from avian species. Influenza A...
Mortality associated with influenza and respiratory syncytial virus in the United States
Thompson WW, et al
Journal of the American Medical Association, 289, 179-186 (2003)

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。