SAB3500976

Sigma-Aldrich

Anti-TMEM107 antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

分子量

predicted mol wt 16 kDa

species reactivity

human

濃度

1 mg/mL

アプリケーション

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

Gene Information

human ... TMEM107(84314)

免疫原

Antibody was raised against a 19 amino acid peptide near the amino terminus of human TMEM107.

特徴および利点

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

関連事項

The action of this antibody can be blocked using blocking peptide SBP3500976.

物理的形状

1 mg/mLのPBS溶液(0.02%アジ化ナトリウム含有)

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Karen S Ho et al.
Current opinion in neurobiology, 12(1), 57-63 (2002-02-28)
Signaling by Sonic hedgehog (Shh) controls important developmental processes, including dorsoventral neural tube patterning, neural stem cell proliferation, and neuronal and glial cell survival. Shh signaling involves lipid modifications to Shh itself, as well as changes in protein subcellular localization....
Kasey J Christopher et al.
Developmental biology, 368(2), 382-392 (2012-06-16)
Cilia are dynamic organelles that are essential for a vast array of developmental patterning events, including left-right specification, skeletal formation, neural development, and organogenesis. Despite recent advances in understanding cilia form and function, many key ciliogenesis components have yet to...
Hilary F Clark et al.
Genome research, 13(10), 2265-2270 (2003-09-17)
A large-scale effort, termed the Secreted Protein Discovery Initiative (SPDI), was undertaken to identify novel secreted and transmembrane proteins. In the first of several approaches, a biological signal sequence trap in yeast cells was utilized to identify cDNA clones encoding...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。