SAB3501008

Sigma-Aldrich

抗ZFX抗体 ウサギ宿主抗体

affinity isolated antibody

NACRES:
NA.41

由来生物

rabbit

品質水準

100

抗体製品の状態

affinity isolated antibody

antibody product type

primary antibodies

クローン

polyclonal

形態

buffered aqueous solution

species reactivity

mouse, human, rat

濃度

1 mg/mL

アプリケーション

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

結合体

unconjugated

NCBIアクセッション番号

UniProtアクセッション番号

出荷済み

wet ice

保管温度

−20°C

免疫原

Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ZFX.

特徴および利点

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

関連事項

The action of this antibody can be blocked using blocking peptide SBP3501008.

物理的形状

1 mg/mLのPBS溶液(0.02%アジ化ナトリウム含有)

免責事項

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

引火点(°F)

Not applicable

引火点(℃)

Not applicable

試験成績書(COA)
原産地証明書(COO)
Jose M Galan-Caridad et al.
Cell, 129(2), 345-357 (2007-04-24)
Stem cells (SC) exhibit a unique capacity for self-renewal in an undifferentiated state. It is unclear whether the self-renewal of pluripotent embryonic SC (ESC) and of tissue-specific adult SC such as hematopoietic SC (HSC) is controlled by common mechanisms. The...
A Schneider-Gädicke et al.
Nature, 342(6250), 708-711 (1989-12-07)
The ZFY gene in the sex-determining region of the human Y chromosome encodes a protein with 13 zinc fingers, and may determine whether an embryo develops as a male or female. ZFX, a related gene on the human X chromosome...
Teresita L Arenzana et al.
Blood, 113(23), 5857-5867 (2009-03-31)
The development, homeostasis, and function of B lymphocytes involve multiple rounds of B-cell receptor (BCR)-controlled proliferation and prolonged maintenance. We analyzed the role of transcription factor Zfx, a recently identified regulator of hematopoietic stem cell maintenance, in B-cell development and...

ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.

テクニカルサービス連絡先

ソーシャルメディア

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

研究、開発、製造

私たちは、バイオテクノロジーと医薬品の研究&開発および製造のソリューションとサービスを提供する、グローバルなライフサイエンス業界のリーディングサプライヤーです。

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

当ウェブサイトのいかなる内容についても、許可なく複写することを禁じます。