1 of 4
当該品目 | Z325333 | Z325104 | Z325287 |
|---|---|---|---|
| capacity 10 mL | capacity 10 mL | capacity 100 mL | capacity 0.5 mL |
| feature class AS | feature class AS | feature class AS | feature class AS |
| manufacturer/tradename BRAND 29712 | manufacturer/tradename BRAND 29732 | manufacturer/tradename BRAND 29719 | manufacturer/tradename BRAND 29721 |
| material AR-Glas® | material AR-Glas® | material AR-Glas® | material AR-Glas® |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 12 ea |
| description total delivery Ex + 15 sec waiting time | description partial delivery | description total delivery Ex + 15 sec waiting time | description partial delivery |
詳細
全BLAUBRANDホールピペットにはバッチ番号が付けられ、オリジナル包装にはバッチ証明が添付されています。
DIN EN ISO 648に準拠
DIN EN ISO 648に準拠
その他情報
BR29712 (India)
法的情報
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
プロトコル
This page contains detailed information on working with and handling pipettes and pipetting aids.