サイズを選択してください
6 EA
¥24,900
¥24,900
カートの在庫状況を確認する
この商品について
NACRES:
NB.01
UNSPSC Code:
41121500
Capacity:
25 mL
Material:
AR-Glas®
Feature:
class AS
Packaging:
pack of 6 ea
スキップする
material
AR-Glas®
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29715
capacity
25 mL
Quality Level
類似した製品をお探しですか? 訪問 製品比較ガイド
1 of 4
当該品目 | Z325376 | Z324930 | Z325325 |
|---|---|---|---|
| material AR-Glas® | material AR-Glas® | material AR-Glas® | material AR-Glas® |
| feature class AS | feature class AS | feature class AS | feature class AS |
| manufacturer/tradename BRAND 29715 | manufacturer/tradename BRAND 29735 | manufacturer/tradename BRAND 29704 | manufacturer/tradename BRAND 29727 |
| capacity 25 mL | capacity 25 mL | capacity 2.5 mL | capacity 5 mL |
| packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea | packaging pack of 6 ea |
| description total delivery Ex + 15 sec waiting time | description partial delivery | description total delivery Ex + 15 sec waiting time | description partial delivery |
General description
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
According to DIN EN ISO 648.
Other Notes
BR29715 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
AR-Glas is a registered trademark of Schott AG
プロトコル
This page contains detailed information on working with and handling pipettes and pipetting aids.