Merck
All Photos(1)
Z325074
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 30 mL, one mark
NACRES:
NB.01
物質
AR-Glas®
詳細
total delivery Ex + 15 sec waiting time
特徴
class AS
包装
pack of 6 ea
manufacturer/tradename
BRAND 29716
キャパシティ
30 mL
Looking for similar products? Visit Product Comparison Guide
Compare Similar Items
View Full Comparison
1 of 4
This Item
Z325066Z325082Z325090
material
AR-Glas®
material
AR-Glas®
material
AR-Glas®
material
AR-Glas®
feature
class AS
feature
class AS
feature
class AS
feature
class AS
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29716
manufacturer/tradename
BRAND 29715
manufacturer/tradename
BRAND 29717
manufacturer/tradename
BRAND 29718
capacity
30 mL
capacity
25 mL
capacity
40 mL
capacity
50 mL
description
total delivery Ex + 15 sec waiting time
description
total delivery Ex + 15 sec waiting time
description
total delivery Ex + 15 sec waiting time
description
total delivery Ex + 15 sec waiting time
詳細
Additional to DIN
全BLAUBRANDホールピペットにはバッチ番号が付けられ、オリジナル包装にはバッチ証明が添付されています。
DIN EN ISO 648に準拠
その他情報
BR29716 (India)
法的情報
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service