スキップする
Z326399
BRAND® BLAUBRAND® volumetric flask, PP stopper, amber glass
volume 10 mL, accuracy: 0.04 mL
別名:
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
ログインで組織・契約価格をご覧ください。
サイズを選択してください
表示を変更する
サイズ/SKU在庫状況単価
2 ea
カートの在庫状況を確認する
¥14,200
この商品について
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
10 mL
Material:
borosilicate glass 3.3 flask, wide-neck
Packaging:
pack of 2 ea
¥14,200
カートの在庫状況を確認する
​
テクニカルサービス
お困りのことがあれば、経験豊富なテクニカルサービスチームがお客様をサポートします。
お手伝いします
material
borosilicate glass 3.3 flask, wide-neck
joint
joint ST/NS neck10/19
description
calibrated to contain (TC, IN)
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37402
volume measuring range
10 mL, accuracy: 0.04 mL
capacity
10 mL
General description
The BLAUBRAND® volumetric flask combines precision and reliability, featuring a square grip PP stopper for easy handling. Crafted from high-quality Boro 3.3 glass and calibrated to contain (TC, In), each flask is marked with high-contrast white enamel for clear readability. Supplied with a batch number and certificate, these flasks ensure quality and traceability, making them ideal for laboratory use.
Other Notes
BR37402 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
類似品目との比較
比較内容をすべて表示
相違点を表示
1 of 1
当該品目
Z326380Z326402Z326437
description
volume 10 mL, accuracy: 0.04 mL
description
volume 5 mL, accuracy: 0.04 mL
description
volume 20 mL, accuracy: 0.04 mL
description
volume 50 mL, accuracy: 0.08 mL
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
capacity
10 mL
capacity
5 mL
capacity
20 mL
capacity
50 mL
manufacturer/tradename
BRAND 37402
manufacturer/tradename
BRAND 37401
manufacturer/tradename
BRAND 37403
manufacturer/tradename
BRAND 37406
joint
joint ST/NS neck10/19
joint
joint ST/NS neck10/19
joint
joint ST/NS neck10/19
joint
joint ST/NS neck14/23
material
wide-neck , borosilicate glass 3.3 flask
material
wide-neck , borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask
material
borosilicate glass 3.3 flask, wide-neck
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
Still not finding the right product?
最新バージョンのいずれかを選択してください:
試験成績書(COA)