natural rubber
3 valves
for pipettes up to 100 mL
BRAND 25315
100 mL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | Z333972 | Z333980 | Z740086 |
---|---|---|---|
material natural rubber | material natural rubber | material natural rubber | material colorless , colorless polypropylene tip |
manufacturer/tradename BRAND 25315 | manufacturer/tradename BRAND 25400 | manufacturer/tradename BRAND 25300 | manufacturer/tradename BRAND 702600 |
volume 100 mL | volume 100 mL | volume 10 mL | volume 0.5-5 mL |
description 3 valves | description 2 valves | description 3 valves | description - |
feature for pipettes up to 100 mL | feature FLIP, for pipettes up to 100 mL | feature Standard, for pipettes up to 10 mL | feature bulk, XXL |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service