Merck
All Photos(1)
Z369101
Finnpipette® Stepper pipette tips
maximum volume 5 mL
NACRES:
NB.01
物質
colorless
無菌性
non-sterile
特徴
barrier: no
包装
pack of 50 ea
manufacturer/tradename
Finn 9404200
最大容量
5 mL
Looking for similar products? Visit Product Comparison Guide
Compare Similar Items
View Full Comparison
1 of 4
This Item
Z369144Z369063Z369071
material
colorless
material
colorless
material
colorless
material
colorless
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
feature
barrier: no
feature
barrier: no
feature
barrier: no
feature
barrier: no
packaging
pack of 50 ea
packaging
pack of 10 ea
packaging
pack of 100 ea
packaging
pack of 100 ea
manufacturer/tradename
Finn 9404200
manufacturer/tradename
Finn 9404230
manufacturer/tradename
Finn 9404170
manufacturer/tradename
Finn 9404180
maximum volume
5 mL
maximum volume
50 mL
maximum volume
0.5 mL
maximum volume
1.25 mL
詳細
Delivers 100-500 μl
Seven sizes, each available in bulk pack or sterile single-wrapped.
包装
Bulk pack
その他情報
Browse all pipette tips
法的情報
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).
Example:
T1503
Product Number
-
25G
Pack Size/Quantity
Additional examples:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
(enter as 1.000309185)
Having trouble? Feel free to contact Technical Service for assistance.
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich Products
  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').
  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').
  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.
Not Finding What You Are Looking For?
In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.
Request COA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service