colorless
sterile
barrier: no
pack of 25 ea
Finn 9404203
5 mL
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | Z369101 | Z369063 | Z369152 |
---|---|---|---|
material colorless | material colorless | material colorless | material colorless |
sterility sterile | sterility non-sterile | sterility non-sterile | sterility sterile |
feature barrier: no | feature barrier: no | feature barrier: no | feature barrier: no |
packaging pack of 25 ea | packaging pack of 50 ea | packaging pack of 100 ea | packaging pack of 50 ea |
manufacturer/tradename Finn 9404203 | manufacturer/tradename Finn 9404200 | manufacturer/tradename Finn 9404170 | manufacturer/tradename Finn 9404173 |
maximum volume 5 mL | maximum volume 5 mL | maximum volume 0.5 mL | maximum volume 0.5 mL |
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service