polystyrene (with 100,000 D MWCO regenerated cellulose semi-permeable membrane)
Equivelent to 16 T-225 standard flasks
sterile; γ-irradiated
Production of monoclonal antibodies and cellular proteins
case of 5
Cole Parmer 2440654
350 mL
類似した製品をお探しですか? Visit 製品比較ガイド
1 of 4
当該品目 | DWKWCL0350-1 | Z688045 | DWKWCL1000-3 |
---|---|---|---|
sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
packaging case of 5 | packaging case of 1 ea | packaging case of 3 | packaging case of 3 ea |
manufacturer/tradename Cole Parmer 2440654 | manufacturer/tradename WHEATON® WCL0350-1 | manufacturer/tradename Cole Parmer 2440655 | manufacturer/tradename WHEATON® WCL1000-3 |
capacity 350 mL | capacity - | capacity 1000 mL | capacity - |
description Equivelent to 16 T-225 standard flasks | description - | description - | description - |
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
製品番号の調べ方
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください (例 T1503).
例
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
COAのロット番号・バッチ番号の調べ方
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
テクニカルサービス連絡先