Merck
Z688045
CELLine 付着性バイオリアクターフラスコ
AD 1000 (For adherent cell types)
NACRES:
NB.22
物質
polystyrene (with 100,000 D MWCO regenerated cellulose semi-permeable membrane)
polystyrene
無菌性
sterile; γ-irradiated
包装
case of 3
manufacturer/tradename
Cole Parmer 2440655
キャパシティ
1000 mL
関連するカテゴリー
類似品目との比較
比較内容をすべて表示
Show Differences
1 of 4
当該品目
Z688037PFHYS1008DWKWCL1000-3
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
packaging
case of 3
packaging
case of 5
packaging
pack of 8
packaging
case of 3 ea
manufacturer/tradename
Cole Parmer 2440655
manufacturer/tradename
Cole Parmer 2440654
manufacturer/tradename
Millicell®
manufacturer/tradename
WHEATON® WCL1000-3
capacity
1000 mL
capacity
350 mL
capacity
-
capacity
-
詳細
Ideal for MAb and cellular protein production. Viable in vitro alternative to using mouse ascites. Bioreactors will utilize innovative membrane technology to separate the cell cultivation area from the media chamber. Cell and cellular-secreted product with a MWCO of 10 kDa are retained between two non-cytotoxic membranes. The upper semi-permeable membrane allows continuous nutrient diffusion and waste elimination. The lower membrane provides direct oxygenation and ensures optimum gas exchange.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
ウェブサイトの表示では、製品番号に容量/数量が連結しています(例:T1503-25G)。確認してください 製品番号欄には製品番号のみを入力してください ( T1503).
T1503
製品番号
-
25G
容量/数量
その他の例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
入力例:1.000309185)
ご不明な点がございましたら、お気軽にお問い合わせくださいテクニカルサービス お問い合わせ
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich製品
  • もしTO09019TOなどのロット番号を見つけた場合は、最初の2文字を除いたロット番号09019TOを入力します。
  • もし05427ES-021などの充てんコード付きロット番号を見つけた場合は、充てんコードの-021を除いたロット番号05427ESを入力します。
  • もしSTBB0728K9などの充てんコード付きロット番号を見つけた場合は、充てんコードのK9を除いたロット番号STBB0728を入力します。
お探しのものが見つかりませんか?
オンラインでCOAを入手できない場合があります。検索してもCOAが見つからない場合は、リクエストできます。
COAをリクエストする
この製品を見ている人はこちらもチェック
テクニカルサービス連絡先