表示を変更する
| 容量 | カタログ番号 | 在庫状況 | 単価 |
|---|
この商品について
UNSPSC Code:
41121800
Material:
clear , flat bottom, polyactide (PLA)
Sterility:
sterile
Feature:
wells 96, lid: no
現在、価格および在庫状況を閲覧できません。
material
clear , flat bottom, polyactide (PLA)
sterility
sterile
feature
wells 96, lid: no
packaging
pack of 100 ea (20 bags of 5 plates)
manufacturer/tradename
Diversified Biotech PLATE-1000
類似した製品をお探しですか? 訪問 製品比較ガイド
General description
100% Plant-Based Planet-Safe® 96-Well Flat bottom Microplates are made from PLA, a plant-based resin derived from non-GMO sustainably cultivated sugarcane. PLA is considerably more hydrophilic than petrochemical resins such as polystyrene. This beneficial property allows a variety of anchorage-dependent eukaryotic cell species to grow in liquid culture on the dish surface without added coatings or treatments. E-Beam Sterilized.
Application
Cell Culture
Features and Benefits
Derived from 100% renewable resources.
Legal Information
Planet-Safe is a registered trademark of Diversified Biotech, Inc.
グローバルトレードアイテム番号
| カタログ番号 | GTIN |
|---|---|
| Z744315-100EA | 04065273747423 |