製品名
Anti-MTMR2 (N-term) antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution
製品番号
SAB1300358
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Appearancecolorless liquid
Concentration>= 0.25 mg/ml
ELISA1:1000
Western Blotting1:100-1:500
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified