製品名
ANTI-RENT1 (N-TERM E22) antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
製品番号
SAB1305873
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
AppearanceReport Result
ConcentrationReport Result
ImmunohistochemistryNot determined
Western Blotting ( Cell Lysate)1:1000
Flow CytometryNot determined
ELISANot determined
ImmunoprecipitationNot determined
Chromatin ImmunoprecipitationNot determined
ImmunofluorescenceNot determined