製品名
ANTI-PSTPIP1(N-TERMINAL) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
製品番号
SAB1306951
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
AppearanceReport Result
ConcentrationReport Result
ImmunohistochemistryNot determined
Western Blotting ( Cell Lysate)1:1000
Flow Cytometry1:10~50
ELISANot determined
ImmunoprecipitationNot determined
Chromatin ImmunoprecipitationNot determined
ImmunofluorescenceNot determined