製品名
Anti-AQP7 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
製品番号
SAB1401014
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Western Blotting (Cell lysate)Antibody concentration: 1 - 5 ug/ml. Specific band of ~37.23 kDa using Jurkat lysate.
Western Blotting (Transfected lysate)Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~37.20 kDa using AQP7 transfected 293T lysate.
Detection AntibodySuitable as a detection antibody in sandwich ELISA assay.