製品名
抗PRSS1抗体 ウサギ宿主抗体 purified immunoglobulin, buffered aqueous solution
製品番号
SAB1401315
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Western Blotting (Tissue lysate)Antibody concentration: 1 - 5 ug/ml. Specific band of ~26.56 kDa using human pancreas lysate.
Western Blotting (Transfected lysate)Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~26.60 kDa using PRSS1 transfected 293T lysate.
Detection AntibodySuitable as a detection antibody in sandwich ELISA assay.