製品名
Anti-ZNF266 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
製品番号
SAB1401536
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Western Blotting (Transfected lysate)Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~29.90 kDa using ZNF266 transfected 293T lysate.
Detection AntibodySuitable as a detection antibody in sandwich ELISA assay.