製品名
Anti-ELAC2 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
製品番号
SAB1401792
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Western Blotting (Tissue lysate)Antibody concentration: 1 - 5 ug/ml. Specific band of ~92.22 kDa using human liver lysate.
Western Blotting (Transfected lysate)Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~92.20 kDa using ELAC2 transfected 293T lysate.
Detection AntibodySuitable as a detection antibody in sandwich ELISA assay.