製品名
ANTI-PIM1 antibody produced in mouse clone 3D12, purified immunoglobulin, buffered aqueous solution
製品番号
SAB1411966
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Appearancecolorless liquid
ConcentrationReport Result
Storage/StabilityReport Result
Storage Temperature-20�C
Species ReactivityReport Result
ELISAAntibody concentration: 1 ug/ml using recombinant protein.
Sandwich ELISADetection limit for recombinant GST tagged PIM1 is approximately 30ng/ml.
Western Blotting (Recombinant Protein)Antibody concentration: 1 ug/ml. Specific band of ~36.7 kDa using immunogen protein lysate.
Western Blotting (Cell Lysate)Not Verified
Western Blotting (Tissue lysate)Not Verified
Western Blotting (Transfected lysate)Not Verified
ImmunohistochemistryNot Verified
ImmunofluorescenceAntibody concentration: 10 ug/ml using HeLa cell.
HandlingReport Result