製品名
Anti-PEPD antibody produced in rabbit affinity isolated antibody
製品番号
SAB2103122
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
Concentration0.5-1 mg/ml
ELISANot Verified
Western BlottingAntibody dilution: 1 µg/ml
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified