製品名
Anti-NANP (AB2) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
製品番号
SAB2900072
製品ブランド
SIGMA
CAS番号
分子量
試験
仕様
AppearanceLiquid
Concentration1 mg/ml
ELISANot Tested
Western BlottingNot Tested
Immunohistochemistry-ParaffinAntibody Concentration: (10-15 ug/mL)
ImmunofluorescenceNot Tested
ImmunoprecipitationNot Tested
Flow CytometryNot Tested
Chromatin ImmunoprecipitationNot Tested